Learn More
Bioworld L-Cystine, 1 kg
Supplier:  Bioworld 403001442
Encompass_Preferred
A non-essential amino acid vital for human development.Synonym: (R,R)-3,3′-Dithiobis(2-aminopropionicacid) Appearance: White crystalline powderDensity : 1.68 g/cm3HS Code : 29309014IUPAC_Name: (2R)-2-Amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acidMelting Point: >240 °CRTECS Number: HA2690000
Catalog No. 50-255-691