Learn More
Research Products International Corp L-Cystine 5 KG

Supplier: Research Products International Corp C810605000.0
CAS Number: 56-89-3
Molecular Formula: C6H12N2O4S2
Molecular Weight: 240.3
Chemical Purity: >98.5%
Soluble in:?1M HCl
Store at:?Room Temperature
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.