Learn More
Medchemexpress LLC L-Cystine-d4 | 1192736-38-1 | 244.33 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN0394S15MG
Encompass_Preferred
L-Cystine-d4 is the deuterium labeled L-Cystine, an amino acid and intracellular thiol crucial for regulating cellular processes. This compound can be used as a tracer and an internal standard for quantitative analysis by techniques like NMR, GC-MS, or LC-MS.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Applicable in NMR, GC-MS, or LC-MS
Catalog No. 50-003-89401