Learn More
Zymo Research Corporation L-Desulphonation Buffer (80 ml)
SDP
Supplier:  Zymo Research Corporation D50465
Encompass_Preferred
Used for in-column desulphonation of bisulfite converted DNA in the EZ DNA Methylation - Lightning kits.
Catalog No. 50-203-7132