Learn More
Medchemexpress LLC L-dithiothreitol | 16096-97-2 | MFCD00064305 | 98.0% | 154.25 g·mol⁻¹ | C4H10O2S2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15917A10MG
Encompass_Preferred
L-dithiothreitol (DTT) is a small-molecule thiol reducing agent used to cleave disulfide bonds and maintain cysteine residues in the reduced state during biochemical and molecular biology workflows. It is commonly used in sample preparation, protein analysis, and enzymatic assays.
  • Effective reducing agent for cleaving disulfide bonds.
  • Helps maintain proteins in a reduced, active state.
  • Soluble in water and compatible with common buffers.
  • High purity suitable for biochemical applications.
  • Available in small quantities for analytical and research use.
Catalog No. 50-002-88109