Learn More
Enzo Life Sciences L-Ergothioneine (10mg). CAS: 497-30-3
SDP
Supplier:  Enzo Life Sciences BMLFR1110010
Encompass_Preferred
Naturally occuring ubiquitous antioxidant which is found in many cells at concentrations up to 2 mM. Peroxynitrate ONOO- scavenger. Protects human neuronal hybridoma cell line (N-18-RE-105) from ONOO--induced oxidative damage. Purity: ≥98% (TLC). Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-0832