Learn More
Cayman Chemical L-ErgothIonIn 25mg
SDP
Supplier:  Cayman Chemical 1490525 MG
Encompass_Preferred
A naturally-occurring amino acid which is a stable antioxidant; scavenges free radicals and oxidants, controls NF-κB activation, and inhibits inflammatory gene expression; blocks oxidative DNA damage and cell death
Catalog No. 50-296-9310