Learn More
Cayman Chemical L-Fucose 1g
SDP
Supplier:  Cayman Chemical 164791 G
Encompass_Preferred
A deoxyhexose monosaccharide found on N- and O-linked glycans and glycolipids that exists either as a terminal modification of glycan structures or serves as a point of attachment for adding other sugars; plays a role in A and B blood group antigen substructure determination, selectin-mediated leukocyte-endothelial adhesion, and host-microbe interactions
Catalog No. 50-296-9871