Learn More
Medchemexpress LLC L-galactono-1,4-lactone | 1668-08-2 | MFCD00066466 | 98.0% | 178.14 g/mol | C6H10O6 | 100 MG

Supplier: Medchemexpress LLC HYW155493100MG
L-galactono-1,4-lactone is a white to off-white solid biochemical reagent used in laboratory research, notably as an intermediate in ascorbate (vitamin C) biosynthesis and in enzymatic assays. For research use only; not for human therapeutic use.
- Purity 98.0%.
- Molecular weight 178.14 g/mol.
- Available in multiple pack sizes (5 mg, 10 mg, 50 mg, 100 mg, 200 mg, 500 mg).
- Storage: room temperature stability for 3 years; in solvent -80°C for 2 years, -20°C for 1 year.
- Appearance white to off-white solid.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.