Learn More
Medchemexpress LLC L-Galactose | 15572-79-9 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN6614
Encompass_Preferred
L-Galactose is a key intermediate in the de novo synthesis of vitamin C (L-ascorbic acid) in plants. It is converted into L-ascorbic acid by participating in the VTC2 cycle and enzymatic steps, exerting antioxidant activities, maintaining cell structure, and participating in the carbon flow distribution of photosynthesis. It can be used to study the analysis of the synthesis pathway of vitamin C in plant physiology.
  • Key intermediate in plant vitamin C synthesis.
  • Converted into L-ascorbic acid.
  • Exhibits antioxidant activities.
  • Maintains cell structure and supports carbon flow in photosynthesis.
  • Useful for studying vitamin C synthesis pathways in plants.
  • Acts as a substrate for L-Galactose dehydrogenase in enzymatic reactions.
  • Functions as a human endogenous metabolite.
Catalog No. 50-004-21860