Learn More
Medchemexpress LLC L-Glucose-13C | 478519-02-7 | 98.0% | 181.15 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW010042S10MG
Encompass_Preferred
L-Glucose-13C is the ⁴³C labeled L-Glucose, which is an enantiomer of D-glucose and can promote food intake. This compound is intended for research use only and not for patient consumption. It can be utilized as a tracer and an internal standard in quantitative analysis.
  • Can be used as a tracer and internal standard
  • Suitable for quantitative analysis by NMR, GC-MS, or LC-MS
  • ⁴³C labeled L-Glucose
  • Enantiomer of D-glucose
  • Promotes food intake
Catalog No. 50-004-53977