Learn More
Abcam L-Glutamate, e x citatory neurotransmitter, 1G
Supplier:  Abcam AB1200491G
Encompass_Preferred
MW 147.13 Da, Purity >99%. Major excitatory neurotransmitter. Acts at glutamate receptors. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1207411