Learn More
Medchemexpress LLC L-Glutamic-2,3,3,4,4-d5 acid | 2784-50-1 | 99.8% | 152.16 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14608S710MM/1ML
Encompass_Preferred
L-Glutamic acid-d5 is a deuterium labeled form of L-Glutamic acid. L-Glutamic acid functions as an excitatory transmitter and an agonist at all subtypes of glutamate receptors, including metabotropic, kainate, NMDA, and AMPA receptors. It also directly activates the release of DA from dopaminergic terminals. This product is for research use only and not intended for patient use.
  • Deuterium labeled L-Glutamic acid
  • Acts as an excitatory transmitter
  • Agonist at metabotropic, kainate, NMDA, and AMPA glutamate receptors
  • Directly activates DA release from dopaminergic terminals
  • Can be used as a tracer
  • Suitable as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
Catalog No. 50-003-63096