Learn More
Medchemexpress LLC L-Glutamic acid-1-13C | 81201-99-2 | 99.9% | 148.12 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14608S150MG
Encompass_Preferred
L-Glutamic acid-1-13C is a 13C-labeled L-Glutamic acid that functions as an excitatory transmitter and an agonist at various glutamate receptors, including metabotropic, kainate, NMDA, and AMPA subtypes. It also shows a direct activating effect on the release of dopamine from dopaminergic terminals.
  • Acts as an excitatory transmitter and agonist at glutamate receptors
  • Shows direct activating effect on dopamine release from dopaminergic terminals
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-63093