Learn More
Medchemexpress LLC L-glutamic acid-1-13C | 81201-99-2 | 99.9% | C4(13C)H9NO4 | 10MG

Supplier: Medchemexpress LLC HY14608S110MG
L-glutamic acid-1-13C is the position-1 13C stable isotope-labeled form of L-glutamic acid supplied as a white to off-white solid for research applications. It is intended for use as a tracer and internal standard in NMR, GC-MS, and LC-MS quantitative analyses and metabolic flux studies. Store sealed and protect from moisture and light.
- Position-1 13C stable isotope labeling for tracer studies and internal standard use.
- High purity (99.85%) suitable for analytical applications.
- Soluble in water and DMSO with recommended preparation methods.
- White to off-white solid, easy to handle and weigh.
- Recommended storage: sealed, protect from moisture and light; in solvent, long-term at -80°C.
- For research use only; not for clinical or human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.