Learn More
Medchemexpress LLC L-glutamic acid 1-tert-butyl ester | 45120-30-7 | MFCD00038562 | 97.0% | 203.24 | C9H17NO4 | 5 G

Supplier: Medchemexpress LLC HYW0181545G
H-Glu-OtBu is a glutamic acid-derived linker supplied for research use. It serves as a non-cleavable ADC linker and as an alkyl chain-based PROTAC linker, and is supplied as a powder with formula C9H17NO4 and molecular weight 203.24 g/mol.
- powder physical form for storage and handling
- used as a non-cleavable ADC linker
- used as an alkyl chain-based PROTAC linker
- CAS number 45120-30-7 for precise identification
- molecular weight 203.24 g/mol and formula C9H17NO4
- storage: powder at -20°C for long-term stability
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.