Learn More
Medchemexpress LLC L-Glutamic acid-13C | 115473-51-3 | 99.9% | 148.12 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY14608S1MG
Encompass_Preferred
L-Glutamic acid-13C is the 13C-labeled version of L-Glutamic acid, functioning as an excitatory transmitter and an agonist for various glutamate receptors. It also directly activates the release of dopamine from dopaminergic terminals.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-60116