Learn More
Medchemexpress LLC L-glutamic acid-13C2 | 115473-56-8 | 99.7% | C313C2H9NO4 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14608S1010MG
Encompass_Preferred
L-Glutamic acid-13C2 is the 13C2 isotopically labeled form of L-glutamic acid used as a stable-isotope standard in biochemical research. It is supplied as a dry, sealed solid in research-scale quantities for tracer experiments, mass spectrometry, metabolic labeling, and quantitative studies where isotopic precision and chemical purity are required.
  • Stable 13C2 labeling for tracer and mass spectrometry studies.
  • High chemical purity suitable for analytical applications.
  • Supplied as a dry, sealed powder for long-term storage.
  • Small research-scale pack sizes available, including 10 mg.
  • Accompanying documentation available: COA and SDS.
  • Enables quantitative metabolic and pathway tracing experiments.
Catalog No. 50-002-61591