Learn More
Medchemexpress LLC L-Glutamic acid-13C5,15N | 202468-31-3 | MFCD00144596 | >99.9% | 153.09 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY14608S31MG
Encompass_Preferred
L-Glutamic acid-13C5,15N is a stable isotope-labeled form of L-Glutamic acid, enriched with five carbon-13 and one nitrogen-15 isotopes. It serves as an excitatory transmitter and an agonist at various glutamate receptor subtypes including metabotropic, kainate, NMDA, and AMPA receptors. This compound is valuable as a tracer and internal standard for quantitative analysis in research using techniques like NMR and GC-MS.
  • Stable isotope-labeled L-Glutamic acid
  • Enriched with five carbon-13 and one nitrogen-15 isotopes
  • Acts as an excitatory neurotransmitter
  • Agonist for metabotropic, kainate, NMDA, and AMPA glutamate receptors
  • Suitable for use as a tracer in metabolic studies
  • Can be used as an internal standard for quantitative analysis
  • Ideal for NMR and GC-MS research applications
Catalog No. 50-003-60714