Learn More
Medchemexpress LLC L-glutamic acid, 5-(1,1-dimethylethyl) ester | 2419-56-9 | >98.0% | 203.24 | C9H17NO4 | 100 G

Supplier: Medchemexpress LLC HYW014742100G
(S)-2-Amino-5-(tert-butoxy)-5-oxopentanoic acid is a chiral glutamic acid derivative supplied for research use as a chemical building block and intermediate. It is provided as a white to off-white solid with analytical data confirming identity and high purity suitable for laboratory synthesis and biochemical studies.
- High purity (>98.0% by NMR).
- Chemical formula C9H17NO4; molecular weight 203.24.
- White to off-white solid appearance.
- Stable in powder form at -20°C (3 years) and 4°C (2 years).
- Store in solvent at -80°C (6 months) or -20°C (1 month).
- Intended for use as a synthetic building block in research applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.