Learn More
Medchemexpress LLC L-Glutamic acid | 56-86-0 | MFCD00002620 | ≥98.0% | 147.13 | 10 MM 1 ML
SDP
Supplier:  Medchemexpress LLC HY1460810MM/1ML
Encompass_Preferred
L-Glutamic acid is an excitatory amino acid neurotransmitter that functions as an agonist for various glutamate receptor subtypes including metabolic, NMDA, and AMPA receptors. It also exerts an agonist effect on the release of dopamine (DA) from dopaminergic nerve endings. This compound is valuable for research into neurological diseases.
  • Acts as an excitatory amino acid neurotransmitter.
  • Functions as an agonist for metabolic, NMDA, and AMPA glutamate receptors.
  • Shows an agonist effect on DA release from dopaminergic nerve endings.
  • Useful in the study of neurological diseases.
Catalog No. 50-003-60709