Learn More
Medchemexpress LLC L-glutamic acid (ammonium) | 7558-63-6 | MFCD00039098 | >98.0% | 164.16 g/mol | C5H12N2O4 | 25 G

Supplier: Medchemexpress LLC HYW41970025G
L-Glutamic acid (ammonium) is the monoammonium salt of the amino acid L-glutamic acid, supplied as a research reagent. It functions as an excitatory neurotransmitter analogue and is used in biochemical, pharmacological, and neurobiology studies involving glutamate receptors and signaling pathways. The material is provided as a solid for laboratory assays and reagent preparation.
- Purity greater than 98%.
- Ammonium salt form improves solubility in aqueous solutions.
- Molecular weight 164.16 g/mol for accurate formulation and calculations.
- Suitable for receptor pharmacology and neuronal signaling assays.
- Available in 25 g solid packaging for routine laboratory use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.