Learn More
Medchemexpress LLC L-Glutamic acid-d3 | 203805-84-9 | 99.12% | 150.15 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY14608S81MG
Encompass_Preferred
L-Glutamic acid-d3 is a deuterium-labeled version of L-Glutamic acid. L-Glutamic acid functions as an excitatory neurotransmitter and an agonist for all types of glutamate receptors, including metabotropic, kainate, NMDA, and AMPA receptors. It also directly activates the release of dopamine from dopaminergic terminals.
  • Used as a tracer.
  • Can be utilized as an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes, like deuterium, can impact the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-59825