Learn More
Medchemexpress LLC L-glutamic acid monosodium hydrate | 6106-04-3 | MFCD00150138 | 98.0% | 187.13 | C5H10NNaO5 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW016145100MG
Encompass_Preferred
L-Glutamic acid monosodium hydrate (CAS 6106-04-3) is the monosodium salt and hydrate form of L-glutamic acid used in biochemical research. It is employed as an oxidative stress inducer in experimental models and is provided with supporting documentation to aid reproducible laboratory use.
  • High purity (98.0%) suitable for research applications.
  • Offered in small pack sizes for analytical and preparative work.
  • Water-soluble form for easy dissolution and buffer preparation.
  • Comes with COA, SDS, and datasheet for traceability and compliance.
  • Sodium salt form appropriate for studies requiring the counterion.
  • CAS-registered substance (6106-04-3) for unambiguous identification.
Catalog No. 50-003-02274