Learn More
Medchemexpress LLC L-Glutamic acid, N-(1-oxohexadecyl)-, 5-(2,5-dioxo-1-pyrrolidinyl) ester | 294855-91-7 | 98.0% | 482.61 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY139276
Encompass_Preferred
Pal-Glu(OSu)-OH is a side chain of Liraglutide, which is a glucagon-like peptide-1 (GLP-1) receptor agonist. This product can be used for type 2 diabetes mellitus research.
  • Side chain of Liraglutide
  • Liraglutide is a GLP-1 receptor agonist
  • Can be used for type 2 diabetes mellitus research
  • Binds to the same receptors as endogenous metabolic hormone GLP-1
Catalog No. 50-003-49090