Learn More
Medchemexpress LLC L-Glutamic acid, N-(3-carboxy-1-oxopropyl)-, 1,5-dihexadecyl ester | 125401-63-0 | C41H77NO7 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY150220
Encompass_Preferred
1,5-Dihexadecyl N-(3-carboxy-1-oxopropyl)-L-glutamate is a lipid utilized in the synthesis of phospholipid vesicles. It collaborates with other lipids to transform polydisperse vesicles into uniformly sized vesicles through freeze-thaw and extrusion techniques. This compound has the capability to enhance protein encapsulation efficiency, making it suitable for drug delivery and various biochemical applications.
  • Used in the synthesis of phospholipid vesicles.
  • Collaborates with other lipids to convert polydisperse vesicles into uniformly sized vesicles.
  • Utilizes freeze-thaw and extrusion techniques for vesicle formation.
  • Enhances protein encapsulation efficiency.
  • Suitable for drug delivery.
  • Suitable for biochemical applications.
Catalog No. 50-003-74363