Learn More
Medchemexpress LLC L-Glutamic γ-monohydroxamate | 1955-67-5 | 99.7% | 162.14 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13787450MG
Encompass_Preferred
L-Glutamic γ-monohydroxamate is an antitumor agent that inhibits cell proliferation. It selectively inhibits the uptake of L-histidine into microvascular endothelial cells. As a vanadium ligand, it activates glucose uptake and metabolism, leading to decreased blood glucose levels in vivo.
  • Inhibits cell proliferation
  • Selectively inhibits L-histidine uptake into microvascular endothelial cells
  • Activates glucose uptake and metabolism as a vanadium ligand
  • Leads to decreased blood glucose levels in vivo
  • Antitumor agent
  • Used in L1210 leukemia cells and B16 melanoma studies
Catalog No. 50-003-51209