Learn More
Cayman Chemical L-GlutamIn-d5 5mg
SDP
Supplier:  Cayman Chemical 348395 MG
Encompass_Preferred
An internal standard for the quantification of L-glutamine by GC- or LC-MS
Catalog No. 50-291-1294