Learn More
Medchemexpress LLC L-Glutamine-1-13C | 159663-16-8 | 147.14 | 5 MG

Supplier: Medchemexpress LLC HYN0390S55MG
L-Glutamine-1-13C is the 13C-labeled L-Glutamine. L-Glutamine (L-Glutamic acid 5-amide) is a non-essential amino acid abundantly present throughout the body and involved in many metabolic processes. It provides a source of carbons for oxidation in some cells.
- Can be used as a tracer.
- Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
- Purity: 98.0%.
- Appearance: Solid.
- Color: White to off-white.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.