Learn More
Medchemexpress LLC L-Glutamine-13C5,15N2 | 285978-14-5 | 153.09 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN0390S65MG
Encompass_Preferred
L-Glutamine-13C5,15N2 is a 13C- and 15N-labeled form of L-Glutamine, a non-essential amino acid crucial for many metabolic processes. This compound provides a source of carbons for oxidation in various cells and appears as a white to off-white solid. It is intended for research use only.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-88826