Learn More
Medchemexpress LLC L-glutamine-13C5,15N2,d5 | 2123439-02-9 | 99.9% | 158.13 | 13C5H5D515N2O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN0390S310MG
Encompass_Preferred
L-Glutamine-13C5,15N2,d5 is an isotopically labeled form of the amino acid L-glutamine containing five 13C atoms, two 15N atoms, and five deuterium atoms. Supplied as a 10 mg research pack, it is suitable for analytical and tracer applications such as mass spectrometry and NMR.
  • Isotopically labeled with 13C, 15N, and deuterium for tracer studies.
  • High isotopic and chemical purity for analytical workflows.
  • Small 10 mg pack convenient for method development and limited-sample work.
  • Solid form for accurate weighing and sample preparation.
  • Compatible with mass spectrometry and NMR applications.
Catalog No. 50-002-95837