Learn More
Medchemexpress LLC L-Glutamine-13C5 | 184161-19-1 | 151.11 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN0390S1100MG
Encompass_Preferred
L-Glutamine-13C5 is a 13C-labeled L-Glutamine, a non-essential amino acid abundantly present throughout the body and involved in many metabolic processes. It provides a source of carbons for oxidation in some cells. This compound can be used as a tracer and as an internal standard for quantitative analysis, leveraging stable heavy isotopes for quantitation in drug development processes.
  • Tracer for various applications.
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-88835