Learn More
Medchemexpress LLC L-Glutamine-13C5 | 184161-19-1 | 151.11 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN0390S1
Encompass_Preferred
L-Glutamine-13C5 is the 13C-labeled L-Glutamine. L-Glutamine is a non-essential amino acid abundantly present throughout the body and involved in many metabolic processes, providing a source of carbons for oxidation in some cells. The compound is suitable for various analytical applications.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-89639