Learn More
Medchemexpress LLC L-Glutamine-13C5 | 184161-19-1 | MFCD00144603 | 151.11 | 5 MG
Supplier: Medchemexpress LLC HYN0390S15MG
L-Glutamine-13C5 is the 13C-labeled L-Glutamine, a non-essential amino acid present abundantly throughout the body and involved in many metabolic processes. It provides a source of carbons for oxidation in some cells.
- Can be used as a tracer.
- Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
- Non-essential amino acid.
- Involved in many metabolic processes.
- Provides a source of carbons for oxidation in some cells.