Learn More
Medchemexpress LLC L-Glutamine-15N-1 | 59681-32-2 | 147.14 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN0390S95MG
Encompass_Preferred
L-Glutamine-15N-1 is the 15N-labeled L-Glutamine. L-Glutamine is a non-essential amino acid found abundantly throughout the body and is involved in numerous metabolic processes. It serves as a carbon source for oxidation in certain cells.
  • This compound can be utilized as a tracer.
  • It can also be employed as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process.
  • Deuteration has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-88982