Learn More
Medchemexpress LLC L-Glutamine-15N | 80143-57-3 | MFCD00084215 | 99.6% | 147.14 | C5H10N15NO3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN0390S10MG
Encompass_Preferred
L-Glutamine-15N is the 15N-labeled form of the amino acid L-glutamine, supplied as a solid stable-isotope reagent for metabolic tracing, isotope labeling studies, and mass spectrometry standards.
  • 15N-labeled reagent for metabolic tracing and isotope experiments.
  • High purity suitable for analytical applications (99.6%).
  • Solid physical state for convenient storage and handling.
  • Molecular weight approximately 147.14 g/mol.
  • Available in small research sizes for tracer studies (e.g., 10 mg).
Catalog No. 50-002-96046