Learn More
Medchemexpress LLC L-glutamine-15N2 | 204451-48-9 | MFCD00084216 | 99.1% | 148.13 | C5H10(15N)2O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN0390S810MG
Encompass_Preferred
L-Glutamine-15N2 is the 15N2 isotopically labeled form of the amino acid L-glutamine, supplied as a solid powder for tracer and analytical work. It is intended for use in metabolic flux analysis, NMR, and LC-MS/MS experiments that require 15N labeling.
  • Stable isotope labeled amino acid for tracer and isotope-labeling studies.
  • High chemical purity and isotopic enrichment suitable for LC-MS and NMR workflows.
  • Available in small research pack sizes, including a 10 mg vial.
  • Soluble in water and DMSO with specified solubility and handling instructions.
  • Store powder at -20°C for long-term stability; follow solvent storage recommendations.
Catalog No. 50-002-95846