Learn More
Supply Solutions L-Glutamine, 56-85-9, MFCD00008044, 25g
Supplier:  Supply Solutions SIGMA G854025G
Encompass_Preferred
Linear Formula H2NCOCH2CH2CH(NH2)CO2H, 99.0-101.0%, Molecular Weight 146.14, mp 185 C (dec.) (lit.), meets USP testing specifications, cell culture tested, from non-animal source, Synonym: (S)-2,5-Diamino-5-oxopentanoic acid, L-Glutamic acid 5-amide, Levoglutamide
Catalog No. NC1066515