Learn More
Medchemexpress LLC L-Glutamine-5-13C | 159680-32-7 | 147.14 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN0390S45MG
Encompass_Preferred
L-Glutamine-5-13C is the 13C-labeled L-Glutamine. L-Glutamine (L-Glutamic acid 5-amide) is a non-essential amino acid found abundantly throughout the body and is involved in many metabolic processes. It provides a source of carbons for oxidation in some cells. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during drug development. Deuteration has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes used as tracers for quantitation during drug development.
  • Deuteration can influence pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-88828