Learn More
Medchemexpress LLC L-Glutamine-N2-15N | 80143-57-3 | 147.14 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN0390S100MG
Encompass_Preferred
L-Glutamine-15N is the 15N-labeled L-Glutamine, a non-essential amino acid involved in many metabolic processes. It provides a source of carbons for oxidation in some cells and is suitable for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-88741