Learn More
Medchemexpress LLC L-Glutamine (standard) | 56-85-9 | 146.14 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN0390R50MG
Encompass_Preferred
L-Glutamine (Standard) is the analytical standard of L-Glutamine, intended for research and analytical applications. As a non-essential amino acid abundantly present throughout the body, L-Glutamine is involved in many metabolic processes and provides a source of carbons for oxidation in some cells.
  • Grade of analytical standard, which is the reference standard supplied assay.
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
  • Purity: 98.59%.
  • Provides a source of carbons for oxidation in some cells.
Catalog No. 50-003-89266