Learn More
Medchemexpress LLC L-Glutamine-13C5,15N2,d5 | 2123439-02-9 | 158.13 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN0390S35MG
Encompass_Preferred
L-Glutamine-13C5, 15N2, d5 is the deuterium, 13C-, and 15N-labeled form of L-Glutamine. It is a non-essential amino acid involved in numerous metabolic processes, providing a source of carbons for oxidation in some cells. This product is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-89923