Learn More
Enzo Life Sciences L-Glutathione (oxidized form) (5g). CAS: 27025-41-8
SDP
Supplier:  Enzo Life Sciences ALX157001G005
Encompass_Preferred
Alternative name: GSSG, (gamma-Glu-Cys-Gly)2. Formula: C20H32N6O12S2. MW: 612.6. Purity: ≥95.0% (HPLC). Formulation: Free acid. Solubility: Soluble in water. Long Term Storage: +4°C.
Catalog No. 50-200-7815