Learn More
Medchemexpress LLC L-Hexanoylcarnitine | 22671-29-0 | 99.9% | 259.34 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11314410MG
Encompass_Preferred
L-Hexanoylcarnitine is an acylcarnitine and is found to be associated with celiac disease. It is for research use only and not sold to patients.
  • Endogenous metabolite
  • Associated with celiac disease
  • Disease markers
  • Endocrine diseases
Catalog No. 50-004-37750