Learn More
Medchemexpress LLC L-histidine, 1-[(1,1-dimethylethoxy)carbonyl]-N-[(9H-fluoren-9-ylmethoxy)carbonyl] | 81379-52-4 | MFCD00151928 | 97.6% | 477.51 g/mol | C26H27N3O6 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW02580710G
Encompass_Preferred
Fmoc-His(Boc)-OH is an Fmoc- and Boc-protected histidine derivative used as a protected building block in solid-phase peptide synthesis. The Fmoc group protects the α-amino terminus while the Boc group protects the histidine imidazole side chain, reducing side reactions during coupling and deprotection. Supplied as a solid reagent, it is intended for peptide assembly and research.
  • Protected histidine for solid-phase peptide synthesis.
  • Fmoc N-terminal protection compatible with standard Fmoc-SPPS protocols.
  • Boc side-chain protection minimizes imidazole reactivity during coupling.
  • High purity suitable for research and peptide production.
  • Available in multiple packaging sizes for small- and large-scale synthesis.
Catalog No. 50-003-00126