Learn More
Medchemexpress LLC L-histidine-13C hydrochloride hydrate | 2483735-43-7 | MFCD00047595 | 98.0% | 210.62 g/mol | C513CH12ClN3O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW014423S10MG
Encompass_Preferred
L-histidine-13C hydrochloride hydrate is the 13C-isotopically labeled hydrochloride hydrate salt of the amino acid L-histidine. It is intended for research use as a tracer and as an internal standard in quantitative analytical methods, and is compatible with NMR, GC-MS, and LC-MS workflows.
Catalog No. 50-003-02984