Learn More
AJINOMOTO USA INC L-Histidine (L-His) 1Kg
Supplier:  AJINOMOTO USA INC 32925
Encompass
Chemical Name: (S)-2-Amino-1H-imidazole-4-propionic Acid; Chemical Formula: C6H9N3O2; Molecular Weight: 155.1500; Nitrogen Content: 0.27080000; CAS Number: 71-00-1; Assay: 98.5 to 101.0%; Description: White crystals or crystalline powder; slightly bitter taste 1KG
Catalog No. NC0254989