Learn More
Medchemexpress LLC L-Histidine | 71-00-1 | 155.15 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYN0832500MG
Encompass_Preferred
L-Histidine (Standard) is the analytical standard of L-Histidine, intended for research and analytical applications. It is an essential amino acid for infants and an inhibitor of mitochondrial glutamine transport. It is commonly utilized in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Used as a reference standard in assays
  • Suitable for research and analytical applications
  • Essential amino acid for infants
  • Inhibitor of mitochondrial glutamine transport
Catalog No. 50-004-15578