Learn More
Medchemexpress LLC L-histidine-d5 hydrochloride hydrate | 2483831-75-8 | 99.6% | 214.66 g·mol⁻¹ | C6H7D5ClN3O3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW014423S450MG
Encompass_Preferred
L-histidine-d5 hydrochloride hydrate is the deuterated form of L-histidine provided as the hydrochloride hydrate salt. It is used as an isotopically labeled standard and tracer for analytical workflows, particularly quantitative LC-MS/MS and tracer studies that require mass differentiation.
  • Deuterium-labeled (d5) for reliable mass spectrometry quantitation.
  • Provided as hydrochloride hydrate to enhance solubility and handling.
  • High purity suitable for analytical standards and method development.
  • Available in small to larger pack sizes to support development and testing.
  • Store sealed, away from moisture and light; follow cold storage guidance for solutions.
Catalog No. 50-003-03303