Learn More
AJINOMOTO USA INC L-Histidine Monohydrochloride Monohydrate (L-His HCLH2O) 1Kg
Supplier:  AJINOMOTO USA INC 32929
Encompass
Chemical Name: (2)-2-Amino-1H-imidazole-4-propionic Acid Monohydrochloride Monohydrate; Chemical Formula: C6H9N3O2·HCl·H2O; Molecular Weight: 209.6300; Nitrogen Content: 0.20040000; CAS Number: 5934-29-2; Assay: 98.5 to 101.0%; Description: White crystals or crystalline powder; slightly acid and bitter taste 1kg
Catalog No. NC0533422